Assay Details
| Gene Symbol: DNASE1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038634.1 | NP_001033723.1 | 2 - 3 | 348 | 85 | |
| GenBank mRNA | BC148924.1 | - | 2 - 3 | 354 | 85 | |
| BT029862.1 | - | 2 - 3 | 248 | 85 | ||
| DQ116783.1 | - | 2 - 3 | 348 | 85 | ||
More Information
Set Membership: |
|

DNASE1L1
Genomic Map