Assay Details
Gene Symbol: SPATC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038677.2 | NP_001033766.1 | 2 - 3 | 598 | 55 | |
GenBank mRNA | BC110282.1 | - | 2 - 3 | 599 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
