Assay Details
| Gene Symbol: AICDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038682.3 | NP_001033771.2 | 3 - 4 | 496 | 104 | |
| GenBank mRNA | DQ303466.1 | - | 2 - 3 | 423 | 104 | |
More Information
Set Membership: |
|

AICDA
Genomic Map