Assay Details
| Gene Symbol: ELP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038684.2 | NP_001033773.1 | 7 - 8 | 959 | 88 | |
| GenBank mRNA | BC110206.1 | - | 7 - 8 | 960 | 88 | |
More Information
Set Membership: |
|

ELP4
Genomic Map