Assay Details
| Gene Symbol: RAB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038685.2 | NP_001033774.1 | 3 - 4 | 267 | 58 | |
| GenBank mRNA | BC110198.1 | - | 3 - 4 | 268 | 58 | |
| BC114771.1 | - | 3 - 4 | 338 | 58 | ||
More Information
Set Membership: |
|

RAB4A
Genomic Map