Assay Details
| Gene Symbol: CENPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038686.2 | NP_001033775.1 | 1 - 2 | 71 | 69 | |
| GenBank mRNA | BC110230.1 | - | 1 - 2 | 72 | 69 | |
More Information
Set Membership: |
|

CENPM
Genomic Map