Assay Details
| Gene Symbol: OOSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038687.1 | NP_001033776.1 | 1 - 2 | 109 | 128 | |
| GenBank mRNA | DQ256127.1 | - | 1 - 2 | 109 | 128 | |
More Information
Set Membership: |
|

OOSP2
Genomic Map