Assay Details
| Gene Symbol: OOSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038687.1 | NP_001033776.1 | 4 - 5 | 480 | 80 | |
| NR_024196.1 | 3 - 4 | 298 | 80 | |||
| GenBank mRNA | DQ256127.1 | - | 4 - 5 | 480 | 80 | |
| DQ256128.1 | - | 3 - 4 | 298 | 80 | ||
More Information
Set Membership: |
|

OOSP2
Genomic Map