Assay Details
| Gene Symbol: TIRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039962.1 | NP_001035051.1 | 2 - 3 | 718 | 74 | |
| GenBank mRNA | DQ319072.1 | - | 2 - 3 | 718 | 74 | |
More Information
Set Membership: |
|

TIRAP
Genomic Map