Assay Details
| Gene Symbol: SAA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040505.2 | NP_001035595.1 | 2 - 3 | 293 | 106 | |
| GenBank mRNA | BC109725.1 | - | 2 - 3 | 294 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109725.1, NM_001040505.2 transcripts.
Set Membership: |
|

SAA4
Genomic Map