Assay Details
| Gene Symbol: PRSS58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040509.2 | NP_001035599.1 | 4 - 5 | 898 | 104 | |
| GenBank mRNA | BC109563.1 | - | 4 - 5 | 900 | 104 | |
More Information
Set Membership: |
|

PRSS58
Genomic Map