Assay Details
| Gene Symbol: TM2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040518.2 | NP_001035608.1 | 3 - 4 | 440 | 81 | |
| GenBank mRNA | BC109864.1 | - | 3 - 4 | 441 | 81 | |
More Information
Set Membership: |
|

TM2D3
Genomic Map