Assay Details
| Gene Symbol: RPL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040520.2 | NP_001035610.1 | 4 - 5 | 441 | 74 | |
| GenBank mRNA | BC109810.1 | - | 4 - 5 | 442 | 74 | |
| BT030549.1 | - | 3 - 4 | 417 | 74 | ||
More Information
Set Membership: |
|

RPL7A
Genomic Map