Assay Details
| Gene Symbol: RPL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040520.2 | NP_001035610.1 | 7 - 8 | 715 | 61 | |
| GenBank mRNA | BC109810.1 | - | 7 - 8 | 716 | 61 | |
| BT030549.1 | - | 6 - 7 | 691 | 61 | ||
More Information
Set Membership: |
|

RPL7A
Genomic Map