Assay Details
| Gene Symbol: PRSS37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040569.2 | NP_001035659.1 | 1 - 2 | 32 | 80 | |
| GenBank mRNA | BC109928.1 | - | 1 - 2 | 33 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109928.1, NM_001040569.2 transcripts.
Set Membership: |
|

PRSS37
Genomic Map