Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045901.1 | NP_001039366.1 | 2 - 3 | 258 | 58 | |
| GenBank mRNA | BC112476.1 | - | 2 - 3 | 258 | 58 | |
More Information
Set Membership: |
|

GADD45G
Genomic Map