Assay Details
| Gene Symbol: CD68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045902.1 | NP_001039367.1 | 2 - 3 | 605 | 122 | |
| GenBank mRNA | BC112487.1 | - | 2 - 3 | 605 | 122 | |
More Information
Set Membership: |
|

CD68
Genomic Map