Assay Details
| Gene Symbol: HAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045953.2 | NP_001039418.2 | 3 - 4 | 605 | 132 | |
| GenBank mRNA | BC112670.1 | - | 3 - 4 | 561 | 132 | |
| EV657492.1 | - | 3 - 4 | 612 | 132 | ||
More Information
Set Membership: |
|

HAX1
Genomic Map