Assay Details
| Gene Symbol: ACYP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045982.2 | NP_001039447.1 | 2 - 3 | 177 | 74 | |
| GenBank mRNA | BC114901.1 | - | 2 - 3 | 178 | 74 | |
More Information
Set Membership: |
|

ACYP1
Genomic Map