Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045987.2 | NP_001039452.1 | 2 - 3 | 788 | 94 | |
| GenBank mRNA | BC105364.1 | - | 2 - 3 | 789 | 94 | |
More Information
Set Membership: |
|

GLYCTK
Genomic Map