Assay Details
| Gene Symbol: PNPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046005.2 | NP_001039470.1 | 3 - 4 | 628 | 77 | |
| GenBank mRNA | BC112803.1 | - | 3 - 4 | 629 | 77 | |
| FJ798978.1 | - | 3 - 4 | 491 | 77 | ||
| FJ897536.1 | - | 3 - 4 | 491 | 77 | ||
More Information
Set Membership: |
|

PNPLA2
Genomic Map