Assay Details
| Gene Symbol: GDAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046013.2 | NP_001039478.1 | 7 - 8 | 1039 | 80 | |
| GenBank mRNA | BC112475.1 | - | 7 - 8 | 1040 | 80 | |
More Information
Set Membership: |
|

GDAP2
Genomic Map