Assay Details
| Gene Symbol: IMMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046015.2 | NP_001039480.1 | 13 - 14 | 1656 | 60 | |
| GenBank mRNA | BC111199.1 | - | 13 - 14 | 1657 | 60 | |
More Information
Set Membership: |
|

IMMT
Genomic Map