Assay Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046017.1 | NP_001039482.1 | 2 - 3 | 374 | 114 | |
| GenBank mRNA | BC112548.1 | - | 2 - 3 | 374 | 114 | |
More Information
Set Membership: |
|

SF3B6
Genomic Map