Assay Details
| Gene Symbol: TBXAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046027.2 | NP_001039492.1 | 11 - 12 | 1502 | 60 | |
| GenBank mRNA | BC112647.1 | - | 11 - 12 | 1503 | 60 | |
More Information
Set Membership: |
|

TBXAS1
Genomic Map