Assay Details
| Gene Symbol: ASPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046033.2 | NP_001039498.1 | 4 - 5 | 618 | 120 | |
| GenBank mRNA | BC112799.1 | - | 4 - 5 | 619 | 120 | |
More Information
Set Membership: |
|

ASPA
Genomic Map