Assay Details
| Gene Symbol: ANGPTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046043.2 | NP_001039508.1 | 4 - 5 | 875 | 66 | |
| GenBank mRNA | BC105516.1 | - | 4 - 5 | 875 | 66 | |
| DQ355521.1 | - | 4 - 5 | 740 | 66 | ||
More Information
Set Membership: |
|

ANGPTL4
Genomic Map