Assay Details
| Gene Symbol: FHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046046.2 | NP_001039511.1 | 3 - 4 | 438 | 59 | |
| GenBank mRNA | BC112720.1 | - | 3 - 4 | 439 | 59 | |
More Information
Set Membership: |
|

FHL2
Genomic Map