Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046058.2 | NP_001039523.1 | 10 - 11 | 1115 | 61 | |
| GenBank mRNA | BC112633.1 | - | 10 - 11 | 1116 | 61 | |
| BT030585.1 | - | 11 - 12 | 1130 | 61 | ||
More Information
Set Membership: |
|

PAH
Genomic Map