Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046061.2 | NP_001039526.1 | 1 - 2 | 112 | 83 | |
| GenBank mRNA | BC105397.1 | - | 1 - 2 | 114 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105397.1, NM_001046061.2 transcripts.
Set Membership: |
|

CYCS
Genomic Map