Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046061.2 | NP_001039526.1 | 2 - 3 | 302 | 87 | |
| GenBank mRNA | BC105397.1 | - | 2 - 3 | 304 | 87 | |
More Information
Set Membership: |
|

CYCS
Genomic Map