Assay Details
| Gene Symbol: ARL8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046071.2 | NP_001039536.1 | 3 - 4 | 462 | 72 | |
| GenBank mRNA | BC112814.1 | - | 3 - 4 | 463 | 72 | |
More Information
Set Membership: |
|

ARL8B
Genomic Map