Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046092.2 | NP_001039557.1 | 12 - 13 | 1129 | 121 | |
| GenBank mRNA | BC111361.1 | - | 12 - 13 | 1130 | 121 | |
More Information
Set Membership: |
|

SEPTIN2
Genomic Map