Assay Details
| Gene Symbol: ECSCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046099.2 | NP_001039564.1 | 2 - 3 | 252 | 58 | |
| NM_001244445.1 | NP_001231374.1 | 4 - 5 | 357 | 58 | ||
| NM_001244446.1 | NP_001231375.1 | 3 - 4 | 306 | 58 | ||
| GenBank mRNA | BC112471.1 | - | 2 - 3 | 241 | 58 | |
| DV851687.1 | - | 4 - 5 | 355 | 58 | ||
| EV675062.1 | - | 3 - 4 | 309 | 58 | ||
More Information
Set Membership: |
|

ECSCR
Genomic Map