Assay Details
| Gene Symbol: FAM207A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046108.2 | NP_001039573.1 | 3 - 4 | 378 | 77 | |
| GenBank mRNA | BC112883.1 | - | 3 - 4 | 379 | 77 | |
More Information
Set Membership: |
|

FAM207A
Genomic Map