Assay Details
| Gene Symbol: TIMM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046132.2 | NP_001039597.1 | 2 - 3 | 142 | 99 | |
| GenBank mRNA | BC111687.1 | - | 2 - 3 | 143 | 99 | |
More Information
Set Membership: |
|

TIMM10
Genomic Map