Assay Details
| Gene Symbol: SUN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046165.2 | NP_001039630.1 | 3 - 4 | 306 | 51 | |
| GenBank mRNA | BC110131.1 | - | 3 - 4 | 307 | 51 | |
More Information
Set Membership: |
|

SUN5
Genomic Map