Assay Details
| Gene Symbol: IL1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046210.2 | NP_001039675.1 | 6 - 7 | 848 | 65 | |
| GenBank mRNA | AB219098.1 | - | 5 - 6 | 696 | 65 | |
| BC112665.1 | - | 6 - 7 | 849 | 65 | ||
More Information
Set Membership: |
|

IL1R2
Genomic Map