Assay Details
| Gene Symbol: GOLT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046215.2 | NP_001039680.1 | 1 - 2 | 138 | 122 | |
| GenBank mRNA | BC110261.1 | - | 1 - 2 | 139 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110261.1, NM_001046215.2 transcripts.
Set Membership: |
|

GOLT1B
Genomic Map