Assay Details
| Gene Symbol: GOLT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046215.2 | NP_001039680.1 | 4 - 5 | 490 | 76 | |
| GenBank mRNA | BC110261.1 | - | 4 - 5 | 491 | 76 | |
More Information
Set Membership: |
|

GOLT1B
Genomic Map