Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046252.2 | NP_001039717.1 | 3 - 4 | 436 | 103 | |
| GenBank mRNA | BC112870.1 | - | 3 - 4 | 438 | 103 | |
More Information
Set Membership: |
|

LITAF
Genomic Map