Assay Details
| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046275.2 | NP_001039740.1 | 7 - 8 | 1621 | 101 | |
| GenBank mRNA | BC112797.1 | - | 7 - 8 | 1622 | 101 | |
More Information
Set Membership: |
|

MINDY2
Genomic Map