Assay Details
| Gene Symbol: GEMIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046278.3 | NP_001039743.1 | 1 - 2 | 400 | 73 | |
| GenBank mRNA | BC112855.1 | - | 1 - 2 | 398 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112855.1, NM_001046278.3 transcripts.
Set Membership: |
|

GEMIN6
Genomic Map