Assay Details
| Gene Symbol: GEMIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046278.3 | NP_001039743.1 | 2 - 3 | 537 | 94 | |
| GenBank mRNA | BC112855.1 | - | 2 - 3 | 535 | 94 | |
| DV831538.1 | - | 2 - 3 | 201 | 94 | ||
More Information
Set Membership: |
|

GEMIN6
Genomic Map