Assay Details
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046285.1 | NP_001039750.1 | 4 - 5 | 540 | 77 | |
GenBank mRNA | BC112635.1 | - | 4 - 5 | 540 | 77 | |
BT025423.1 | - | 4 - 5 | 568 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
