Assay Details
| Gene Symbol: NT5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046322.1 | NP_001039787.1 | 2 - 3 | 382 | 62 | |
| GenBank mRNA | BC110222.1 | - | 2 - 3 | 382 | 62 | |
More Information
Set Membership: |
|

NT5C
Genomic Map