Assay Details
| Gene Symbol: SULT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046353.1 | NP_001039818.1 | 4 - 5 | 666 | 96 | |
| GenBank mRNA | BC112644.1 | - | 4 - 5 | 666 | 96 | |
More Information
Set Membership: |
|

SULT2A1
Genomic Map