Assay Details
| Gene Symbol: BUD31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046364.2 | NP_001039829.1 | 1 - 2 | 59 | 92 | |
| GenBank mRNA | BC111638.1 | - | 1 - 2 | 60 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111638.1, NM_001046364.2 transcripts.
Set Membership: |
|

BUD31
Genomic Map