Assay Details
| Gene Symbol: SPINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046367.2 | NP_001039832.1 | 8 - 9 | 1372 | 70 | |
| GenBank mRNA | BC105310.1 | - | 8 - 9 | 1373 | 70 | |
More Information
Set Membership: |
|

SPINT1
Genomic Map