Assay Details
| Gene Symbol: HCFC1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046398.2 | NP_001039863.1 | 1 - 2 | 133 | 132 | |
| GenBank mRNA | BC111671.1 | - | 1 - 2 | 134 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111671.1, NM_001046398.2 transcripts.
Set Membership: |
|

HCFC1R1
Genomic Map